Learn More
Ras homolog enriched in brain, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RHEB protein.
Brand: Abnova H00006009-B01.50uL
Description
This gene is a member of the small GTPase superfamily and encodes a lipid-anchored, cell membrane protein with five repeats of the RAS-related GTP-binding region. This protein is vital in regulation of growth and cell cycle progression due to its role in the insulin/TOR/S6K signaling pathway. The protein has GTPase activity and shuttles between a GDP-bound form and a GTP-bound form, and farnesylation of the protein is required for this activity. Three pseudogenes have been mapped, two on chromosome 10 and one on chromosome 22. [provided by RefSeq
Sequence: MPQSKSRKIAILGYRSVGKSSLTIQFVEGQFVDSYDPTIENTFTKLITVNGQEYHLQLVDTAGQDEYSIFPQTYSIDINGYILVYSVTSIKSFEVIKVIHGKLLDMVGKVQIPIMLVGNKKDLHMERVISYEEGKALAESWNAAFLESSAKENQTAVDVFRRIILEAEKMDGAASQGKSSCSVMSpecifications
| Ras homolog enriched in brain | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RHEB protein. | |
| RHEB | |
| MGC111559/RHEB2 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_005614 | |
| RHEB | |
| RHEB (NP_005605, 1 a.a. ∼ 184 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 6009 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.