missing translation for 'onlineSavingsMsg'
Learn More

rhomboid, veinlet-like 1 (Drosophila), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant RHBDL1.

Brand:  Abnova H00009028-A01.50uL

Product Code. 16179765

  • £209.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MGRVEDGGTTEELEDWDPGTSALPAPGIKQGPREQTGTGPLSQKCWEPEPDAPSQPGPALWSRGRARTQALAGGSSLQQLDPENTGFIGADTFTGLVHSH
Specifications

Specifications

rhomboid, veinlet-like 1 (Drosophila)
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant RHBDL1.
RHBDL1
RHBDL/RRP
Mouse
50 μL
Yes
9028
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
50% glycerol
NM_003961
RHBDL1
RHBDL1 (NP_003952, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.