Learn More
resistin, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RETN protein.
Brand: Abnova H00056729-D01P.100ug
Description
This gene belongs to the family defined by the mouse resistin-like genes. The characteristic feature of this family is the C-terminal stretch of 10 cys residues with identical spacing. The mouse homolog of this protein is secreted by adipocytes, and may be the hormone potentially linking obesity to type II diabetes. [provided by RefSeq
Sequence: MKALCLLLLPVLGLLVSSKTLCSMEEAINERIQEVAGSLIFRAISSIGLECQSVTSRGDLATCPRGFAVTGCTCGSACGSWDVRAETTCHCQCAGMDWTGARCCRVQPSpecifications
| resistin | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human RETN protein. | |
| RETN | |
| ADSF/FIZZ3/MGC126603/MGC126609/RETN1/RSTN/XCP1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56729 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_020415 | |
| RETN | |
| RETN (AAI01555.1, 1 a.a. ∼ 108 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.