missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSV NP Monoclonal Antibody (I25J), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17289
This item is not returnable.
View return policy
Description
The MA1-7289 antibody reacts with Respiratory Sync. virus Np in viral and infected tissue samples. MA1-7289 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7289 antibody was raised against RSV nucleoprotein.
Specifications
| RSV NP | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Immunocytochemistry | |
| I25J | |
| FITC | |
| human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein | |
| RSV nucleoprotein. | |
| 100 μg | |
| Primary | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction