missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Recoverin Monoclonal Antibody (6a55 cd6)
GREENER_CHOICE

Product Code. 11505712
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11505712 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11505712 Supplier Invitrogen™ Supplier No. MA1932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-932 will detect Recoverin from human, rat, bovine, and ovine samples. MA1-932 has been successfully used in Western blot, ICC/IF and IHC (P) procedures. By Western blot, this antibody will detect a ~23 kDa band representing Recoverin from rat brain, as well as in bovine and ovine retina. This antibody is not recommended for rat brain tissue in IHC applications. The MA1-932 immunogen is considered commercially sensitive.

Seems to be implicated in the pathway from retinal rod guanylate cyclase to rhodopsin. May be involved in the inhibition of the phosphorylation of rhodopsin in a calcium-dependent manner. The calcium-bound recoverin prolongs the photoresponse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Recoverin
Applications ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6a55 cd6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene RCVRN
Gene Accession No. P21457, P35243
Gene Alias 23 kDa photoreceptor cell-specific protein; cancer associated retinopathy antigen; cancer associated retinopathy protein; cancer-associated retinopathy protein; CAR; guanylate cyclase activator; p26; P26-calcium binding protein; Protein CAR; RCV1; RCVRN; Recoverin; S-modulin
Gene Symbols RCVRN
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101111206, 140936, 281447, 5957
Target Species Human, Rat, Bovine, Sheep
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.