missing translation for 'onlineSavingsMsg'
Learn More
Learn More
retinol dehydrogenase 11 (all-trans/9-cis/11-cis), Mouse, Clone: 1H6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant RDH11.
Brand: Abnova H00051109-M01.100ug
This item is not returnable.
View return policy
Description
RHD11, a member of the short-chain dehydrogenase/reductase (SDR) superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.[supplied by OMIM
Sequence: IRKMLSSGVCTSTVQLPGKVVVVTGANTGIGKETAKELAQRGARVYLACRDVEKGELVAKEIQTTTGNQQVLVRKLDLSDTKSIRAFAKGFLAEEKHLHVLINNAGVMMCPYSKTADGFEMHIGVNHLGHFLLTHLLLEKLKESAPSRIVNVSSLAHHLGRIHFHNLQGEKFYNAGLAYCHSKLANILFTQELARRLKGSGVTTYSVHPGTVQSELVRHSSFMRWMWWLFSFFIKTPQQGAQTSLHCALTEGLEILSGNHFSDCHVAWVSAQARNETIARRLWDVSCDLLGLPIDSpecifications
| retinol dehydrogenase 11 (all-trans/9-cis/11-cis) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000112 | |
| RDH11 | |
| RDH11 (AAH00112, 24 a.a. ∼ 318 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1H6 | |
| Mouse monoclonal antibody raised against a full length recombinant RDH11. | |
| RDH11 | |
| ARSDR1/CGI-82/FLJ32633/HCBP12/MDT1/PSDR1/RALR1/SCALD/SDR7C1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 51109 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction