Learn More
CGRP receptor component, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RCP9.
Brand: Abnova H00027297-A01.50uL
Description
This gene encodes a membrane protein that functions as part of a receptor complex for a small neuropeptide that increases intracellular cAMP levels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: QQNLNTITYETLKYISKTPCRHQSPEIVREFLTALKSHKLTKAEKLQLLNHRPVTAVEIQLMVEESEERLTEEQIEALLHTVTSILPAEPEAEQKKNTNSNVAMDEEDPASpecifications
| CGRP receptor component | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RCP9. | |
| CRCP | |
| CGRP-RCP/MGC111194/RCP/RCP9 | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_014478 | |
| CRCP | |
| RCP9 (NP_055293, 39 a.a. ∼ 148 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 27297 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.