missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCOR1 Monoclonal Antibody (658526)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524297
This item is not returnable.
View return policy
Description
In direct ELISAs, no cross-reactivity with recombinant human RCOR3 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
RCOR1 is a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression.
Specifications
| RCOR1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9UKL0 | |
| RCOR1 | |
| E. coli-derived recombinant human RCOR1/CoREST Ala394-Ser482. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 658526 | |
| Unconjugated | |
| RCOR1 | |
| 5730409O11; 6720480E22Rik; AU042633; COREST; D12Wsu95e; im:6904264; KIAA0071; LOC102554884; mKIAA0071; Protein CoREST; RCOR; Rcor1; REST corepressor 1; REST corepressor 1-like; Rocr1; si:dkey-46n18.7; zgc:158607 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 23186 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction