missing translation for 'onlineSavingsMsg'
Learn More

OLLAS Epitope Tag Antibody (L2) - BSA Free, Novus Biologicals™

Product Code. p-4984931 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18764983 0.025 mL 25µL
18719493 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18764983 Supplier Novus Biologicals Supplier No. NBP106713SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 14 publications

OLLAS Epitope Tag Monoclonal specifically detects OLLAS Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry Whole-Mount, ChIP assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLLAS Epitope Tag
Applications ChIP Assay, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone L2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:10 - 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10 - 1:100, Immunoblotting reported in scientific literature (PMID 27600537), Immunohistochemistry (Whole-Mount), Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 31175278)
Formulation PBS with 0.05% Sodium Azide
Gene Alias E.coli OmpF Linker and mouse Langerin fusion Sequence, OLLA-2
Host Species Rat
Immunogen OLLAS (SGFANELGPRLMGK)-tagged extracellular domain of mouse Langerin
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Epitope Tags, OLLAS Epitope Tags
Primary or Secondary Primary
Test Specificity Tag specific.
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1 κ
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.