Learn More
Invitrogen™ F(ab')2-Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, PE
F(ab')2-Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ A10544
Product Code. 10428663
  • £354.00 / 250µL
For Research Use Only. All usage must comply with product instructions.
Description
Description
F(ab')2-Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody for Flow
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgG (H+L) Cross-Adsorbed
Polyclonal
PE
Goat
Purified
RUO
Rat
Secondary Antibody
IgG
Flow Cytometry
1 mg/mL
PBS with 5mM sodium azide; pH 7.5
Gamma Immunoglobins Heavy and Light chains.
250 μL
Secondary
4°C