Learn More
Goat anti-Rat IgA Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184707
Code : LD
Additional Details : Weight : 1.24740kg
Description
This antibody may cross-react with IgH from other species. Goat anti Rat IgA antibody recognizes rat IgA as assessed by immunoelectrophoresis and ELISA.Goat anti Rat IgA antibody may cross react with IgA from other species.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgA | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Proprietary Immunogen. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG |