missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Rat CD80/B7-1 (3H5), Proteintech
Mouse Monoclonal Antibody
£37.00 - £109.00
Specifications
| Antigen | CD80/B7-1 |
|---|---|
| Clone | 3H5 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233644
|
Proteintech
65217-1-IG-500UG |
500 μg |
£114.00 £109.00 / 500µg Save £5.00 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236951
|
Proteintech
65217-1-IG-100UG |
100 μg |
£38.00 £37.00 / 100µg Save £1.00 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD80/B7-1 antibody from Proteintech is a mouse monoclonal antibody generated with htlv-1-transformed lewis-s1 rat t cell line of rat CD80/B7-1. This antibody recognizes rat antigen. The CD80/B7-1 antibody has been validated for the following applications: FC analysis.Specifications
| CD80/B7-1 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 25408 | |
| IgG1 κ | |
| Protein G purification | |
| Flow Antibody |
| 3H5 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Rat | |
| HTLV-1-transformed Lewis-S1 rat T cell line | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title