missing translation for 'onlineSavingsMsg'
Learn More

Ras association (RalGDS/AF-6) domain family (N-terminal) member 8, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human RASSF8 protein.

Brand:  Abnova H00011228-D01P.100ug

Product Code. 16153936

  • £316.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MELKVWVDGVQRIVCGVTEVTTCQEVVIALAQAIGRTGRYTLIEKWRDTERHLAPHENPIISLNKWGQYASDVQLILRRTGPSLSERPTSDSVARIPERTLYRQSLPPLAKLRPQIDKSIKRREPKRKSLTFTGGAKGLMDIFGKGKETEFKQKVLNNCKTTADELKKLIRLQTEKLQSIEKQLESNEIEIRFWEQKYNSNLEEEIVRLEQKIKRNDVEIEEEEFWENELQIEQENEKQLKDQLQEIRQKITECENKLKDYLAQIRTMESGLEAEKLQREVQEAQVNEEEVKGKIGKVKGEIDIQGQQSLRLENGIKAVERSLGQATKRLQDKEQELEQLTKELRQVNLQQFIQQTGTKVTVLPAEPIEIEASHADIERGIIILSDKQECKD
Specifications

Specifications

Ras association (RalGDS/AF-6) domain family (N-terminal) member 8
Polyclonal
Rabbit polyclonal antibody raised against a full-length human RASSF8 protein.
RASSF8
C12orf2/HoJ-1
Rabbit
Affinity Purified
RUO
Primary
Human
Antibody
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
NM_007211.2
RASSF8
RASSF8 (NP_009142.2, 1 a.a. ∼ 392 a.a) full-length human protein.
100 μg
Signal Transduction
11228
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.