missing translation for 'onlineSavingsMsg'
Learn More

Ran Mouse, Unlabeled, Clone: 20, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610340

 View more versions of this product

Product Code. 15845409

  • ONLINE EXCLUSIVE PRICE
    £254.00
    £243.04 / 1 set
    Save £10.96
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ran is a highly conserved GTPase that is ubiquitously expressed. At steady-state, 80-90% of cellular Ran is located in the nucleus, with the remainder in the cytoplasm. The human TC4/Ran cDNA was originally isolated based on the predicted homology of its encoded protein product to Ras. Unlike other members of the Ras superfamily, Ran does not appear to contain signals for lipid modification, nor does it appear to show membrane localization. Ran has been implicated in a number of cellular processes such as the initiation of DNA replication, entry into and exit from mitosis and in nuclear RNA and protein transport through the nuclear pore complex. A number of Ran binding proteins have been identified. One of the smaller proteins identified is a 28kDa cytosolic molecule known as RanBP1. The GTP-bound state of Ran is stabilized through its binding to RanBP1.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ran
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG2a
Western Blot
20
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.