missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ral guanine nucleotide dissociation stimulator, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RALGDS protein.
Brand: Abnova H00005900-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MVQRMWAEAAGPAGGAEPLFPGSRRSRSVWDAVRLEVGVPDSCPVVLHSFTQLDPDLPRPESSTQEIGEELINGVIYSISLRKVQLHHGGNKGQRWLGYENESALNLYETCKVRTVKAGTLEKLVEHLVPAFQGSDLSYVTIFLCTYRAFTTTQQVLDLLFKRYGCILPYSDEDGGPQDQLKNAISSILGTWLDQYSEDFCQPPDFPCLKQLVAYVQLNMPGSDLERRAHLLLAQLEHSEPIEAEPEALSPVPALKPTPELELALTPARAPSPVPAPAPEPEPAPTPAPGSELEVAPAPAPELQQAPEPAVGLESAPAPALELEPAPEQDPAPSQTLELEPAPAPVPSLQPSWPSPVVAENGLSEEKPHLLVFPPDLVAEQFTLMDAELFKKVVPYHCLGSIWSQRDKKGKEHLAPTIRATVTQFNSVANCVITTCLGNRSTKAPDRARVVEHWIEVARECRILKNFSSLYAILSALQSNSIHRLKKTWEDVSRDSFRIFQKLSEIFSDENNYSLSRELLIKEGTSKFATLEMNPKRAQKRPKETGIIQGTVPYLGTFLTDLVMLDTAMKDYLYGRLINFEKRRKEFEVIAQIKLLQSACNNYSIAPDEQFGAWFRAVERLSETESYNLSCELEPPSESASNTLRTKKNTAIVKRWSDRQAPSTELSTSGSSHSKSCDQLRCGPYLSSGDIADALSVHSAGSSSSDVEEINISFVPESPDGQEKKFWESASQSSPETSGISSASSSTSSSSASTTPVAATRTHKRSVSGLCNSSSALPLYNQQVGDCCIIRVSLDVDNGNMYKSILVTSQDKAPAVIRKAMDKHNLEEEEPEDYELLQILSDDRKLKIPENANVFYAMNSTANYDFVLKKRTFTKGVKVKHGASSTLPRMKQKGLKIAKGIFSpecifications
| ral guanine nucleotide dissociation stimulator | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human RALGDS protein. | |
| RALGDS | |
| FLJ20922/RGF/RalGEF | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| In situ PLA, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC059362 | |
| RALGDS | |
| RALGDS (AAH59362.1, 1 a.a. ∼ 902 a.a) full-length human protein. | |
| 100 μg | |
| Signal Transduction | |
| 5900 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction