Learn More
RAE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521739
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLaS3, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Pig (100%), Chicken (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RAE1 Polyclonal antibody specifically detects RAE1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
RAE1 | |
Polyclonal | |
Unconjugated | |
RAE1 | |
3230401I12Rik; 41; D2Ertd342e; dJ481F12.3; dJ800J21.1; FLJ30608; HGNC:9828; homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer); MGC117333; MGC126076; MGC126077; MIG14; migration-inducing gene 14; MNRP; Mnrp41; mRNA export factor; mRNA export factor-like protein; mRNA export protein; mRNA-associated protein MRNP 41; mRNA-binding protein, 41-kD; MRNP41; OTTHUMP00000166110; rae1; RAE1 (RNA export 1, S.pombe) homolog; rae1 protein; rae1 protein homolog; RAE1 RNA export 1 homolog; RAE1 RNA export 1 homolog (S. pombe); ribonucleic acid export 1; RP4-800J21.2; zgc:56449; zgc:77723 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
P78406 | |
RAE1 | |
Synthetic peptide corresponding to a region within amino acids 307 and 368 of RAE1 (Uniprot ID#P78406) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |