missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD51 Monoclonal Antibody (51RAD01 (3C10)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514419
This item is not returnable.
View return policy
Description
MA5-14419 targets RAD51 in IHC (P), IP, ICC/IF and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-14419 immunogen is recombinant Rad51 protein.
RAD51 plays a critical role in homologous strand exchange, a key step in DNA repair through homologous recobination. It binds to single and double-stranded DNA and exhibits DNA-dependent ATPase activity. RAD51 catalyzes the recognition of homology and strand exchange between homologous DNA partners to form a joint molecule between a processed DNA break and the repair template. It binds to a single-stranded DNA in an ATP-dependent manner to form nucleoprotein filaments which are essential for the homology search and strand exchange. Mutations in the gene can results in breast cancer, mirror movements 2 and fanconi anemia.
Specifications
| RAD51 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| Q06609, Q08297 | |
| Rad51 | |
| Recombinant Rad51 protein | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 51RAD01 (3C10) | |
| Unconjugated | |
| Rad51 | |
| AV304093; BRCA1/BRCA2-containing complex, subunit 5; BRCC5; DNA repair protein RAD51; DNA repair protein RAD51 homolog 1; DNA repair protein RAD51 homolog A; DNA repair protein rhp51; FANCR; homolog to S.cerevisiae; HRAD51; HsRad51; HsT16930; MGC128559 protein; MRMV2; MUT5; rad51; RAD51 homolog; RAD51 homolog (RecA homolog); RAD51 homolog (RecA homolog, E. coli); RAD51 homolog A; RAD51 recombinase; RAD51 recombinase L homeolog; rad51.1; rad51.L; rad51-1; RAD51A; rad51-a; rad51-b; Reca; RecA family recombinase Rhp51; RecA, E. coli, homolog of; RecA-like protein; recombinase RAD51; recombination protein A; RGD1563603; rhp51; SPAC644.14c; wu:fb38e12; xrad51; XRad51.1; YER095W; zgc:77754 | |
| Mouse | |
| Protein G | |
| RUO | |
| 19361, 499870, 5888 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction