Learn More
Invitrogen™ RAD21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528344
Description
Recommended positive controls: K562, THP-1, HL-60, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (90%), Chicken (94%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.
Specifications
| RAD21 | |
| Polyclonal | |
| Unconjugated | |
| Rad21 | |
| 64-kDa carboxy-terminal product; 64-kDa C-terminal product; 65-kDa carboxy-terminal product; CDLS4; double-strand-break repair protein rad21 homolog; double-strand-break repair protein rad21-like protein; FLJ25655; FLJ40596; h NXP-1; hHR21; HR21; HRAD21; KIAA0078; KIAA0078RAD21; kleisin; MCD1; mHR21; mKIAA0078; nuclear matrix protein 1; NXP1; NXP-1; Pokeweed agglutinin-binding protein 29; protein involved in DNA double-strand break repair; PW29; Rad21; RAD21 cohesin complex component; RAD21 homolog; RAD21 homolog (S. pombe); SCC1; SCC1 homolog; sister chromatid cohesion 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19357, 314949, 5885 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.38 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O60216, Q61550 | |
| Rad21 | |
| Recombinant protein encompassing a sequence within the center region of human Rad21. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.