missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1091
Code : PN
Additional Details : Weight : 0.01000kg
Description
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.Specifications
RAC1 | |
Polyclonal | |
Unconjugated | |
Rac1 | |
AL023026; cell migration-inducing gene 5 protein; D5Ertd559e; GTPase; MGC111543; MIG5; p21-Rac1; Rac 1b; Rac family small GTPase 1; RAC1; Rac-1; Rac2; rac2 GTP-binding protein; ras-like protein TC25; RAS-related C3 botulinum substrate 1; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); rho family, small GTP binding protein Rac1; TC25; TC-25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19353, 5879 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P63000, P63001 | |
Rac1 | |
Recombinant Full-length Human Protein | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |