Learn More
RABGGTB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528324
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, Mouse brain, Rat brain. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (91%), Xenopus laevis (91%), Pig (98%), Sheep (93%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RABGGTB Polyclonal antibody specifically detects RABGGTB in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
RABGGTB | |
Polyclonal | |
Unconjugated | |
RABGGTB | |
fj12b07; geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; Ggtb; rab geranylgeranyl transferase b subunit; Rab geranylgeranyl transferase componenet, subunit beta; Rab geranylgeranyl transferase, b subunit; Rab geranylgeranyltransferase beta subunit; rab geranylgeranyltransferase subunit beta; rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyltransferase, beta subunit; rab GG transferase beta; rab GGTase beta; Rabggtb; Type II protein geranyl-geranyltransferase subunit beta; wu:fj12b07; zgc:56443; zgc:56443 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19352, 25533, 5876 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P53611, P53612, Q08603 | |
RABGGTB | |
Recombinant fragment corresponding to a region within amino acids 37 and 318 of Human RABGGTB | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |