missing translation for 'onlineSavingsMsg'
Learn More

Rabbit IgG Isotype Control, Invitrogen™

Rabbit IgG Isotype Control from Thermo Scientific™

Brand:  Invitrogen PA5-23090

Product Code. 13297738

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Rabbit IgG Isotype Control from Thermo Scientific™

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody’s species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Host Species: Rabbit
Cross Reactivity: Not Applicable
Isotype: IgG
Immunogen: Naive rabbit sera
Formulation: 100μg Purified Isotype Control (0.5mg/ml) in PBS, with 0.05% sodium azide
This product has been successfully used in Control, Flow Cytometry, Immunohistochemistry (Frozen) and Immunohistochemistry (Paraffin) procedures. The immunogen is Naive rabbit sera.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rabbit IgG
Polyclonal
Rabbit
Protein G
Primary
Purified
Control, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Naive rabbit sera
100 μg
Store at -20°C, Avoid Freeze/Thaw Cycles
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.