missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab acceptor 1 (prenylated), Mouse, Clone: 2A4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RABAC1.
Brand: Abnova H00010567-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MAAQKDQQKDAEAEGLSGTTLLPKLIPSGAGREWLERRRATIRPWSTFVDQQRFSRPRNLGELCQRLVRNVEYYQSNSpecifications
| Rab acceptor 1 (prenylated) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006423 | |
| RABAC1 | |
| RABAC1 (NP_006414.1, 1 a.a. ∼ 77 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2A4 | |
| Mouse monoclonal antibody raised against a partial recombinant RABAC1. | |
| RABAC1 | |
| PRA1/PRAF1/YIP3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 10567 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction