Learn More
Invitrogen™ GDI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527498
Description
Recommended positive controls: HepG2, NIH-3T3. Predicted reactivity: Mouse (96%), Rat (97%), Zebrafish (88%), Xenopus laevis (88%), Dog (97%), Rabbit (97%), Sheep (98%), Chimpanzee (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI1 is expressed primarily in neural and sensory tissues. Mutations in GDI1 have been linked to X-linked nonspecific mental retardation.
Specifications
| GDI1 | |
| Polyclonal | |
| Unconjugated | |
| GDI1 | |
| 1A; GDI1; GDI-1; GDIA; GDIalpha; GDIL; GDP dissociation inhibitor 1; GDP-dissociation inhibitor 1; GTP-binding protein; guanosine diphosphate (GDP) dissociation inhibitor 1; guanosine diphosphate dissociation inhibitor 1; mental retardation, X-linked 48; MRX41; MRX48; oligophrenin-2; OPHN2; Protein XAP-4; rab GDI alpha; Rab GDIalpha; rab GDP dissociation inhibitor alpha; rab GDP-dissociation inhibitor, alpha; RABGD RABGDIA; RABGD1A; RABGDIA; SMG p25A GDI; testis secretory sperm-binding protein Li 208a; XAP4; XAP-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14567, 2664 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P31150, P50396 | |
| GDI1 | |
| Recombinant protein encompassing a sequence within the center region of human GDI1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.