Learn More
PU.1 Mouse anti-Human, Unlabeled, Clone: G148-74, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554268
Description
PU.1 is an ets-related transcription factor expressed in B lymphocytes and macrophages. It was initially identified as the oncogene Spi-1 which was found to block erythroblast differentiation. PU.1 functions by binding transcriptional control elements such as μB in the immunoglobulin heavy chain enhancer and π adjacent to μE2. It has been shown to form a complex with NF-EM5, a B cell transcription factor, resulting in the activation of the immunoglobulin κ 3' enhancer. PU.1 also appears to be critically involved in the control of monocyte development by regulating the expression of the macrophage colony-stimulating factor receptor. PU.1 runs as a 40-42kDa protein on SDS-PAGE. G148-74 recognizes human PU.1, an Ets-related transcription factor. PU.1 is expressed in B lymphocytes and macrophages and like other Ets-related proteins, binds to consensus sites in DNA through an 85 amino acid Ets domain in the carboxyl terminal region of the protein. The antibody was raised against a bacterially expressed GST-PU.1 fusion protein.
Host Species: Mouse
Clone: G148-74
Isotype: IgG2a
Species Reactivity: Human
Immunogen: PU.1 Fusion Protein
Formula Weight [Chemical]: 40-42kDa
Immunoprecipitation, Western Blotting
Specifications
| PU.1 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| PU.1 Fusion Protein | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgG2a |
| Western Blot | |
| G148-74 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.