Learn More
PTF1A Mouse, Unlabeled, Clone: S25-763, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564745
Description
The S25-763 monoclonal antibody specifically binds PTF1A (Pancreas specific transcription factor 1a). PTF1A is a transcription factor that contains a basic helix-loop-helix motif domain and, in combination with p75 and p64, forms the pancreas transcription factor complex. PTF1A plays a roles in pancreatic and cerebellar development. In early pancreatic development, pancreatic bud cells can either continue to pancreatic organogenesis or regress to a duodenal fate, and PTF1A plays a role in this differentiation. In late pancreatic development, the expression of PTF1A is restricted to acinar cells where it is involved in the maintenance of exocrine-specific gene expression. PTF1A expression is often evaluated to monitor pancreatic lineage differentiation from pluripotent stem cells. In addition, PTF1A is expressed during retinal development, and mutations in PTF1A cause cerebellar deficiencies.
This mouse anti-human PTF1A monoclonal antibody, clone S25-763, has been demonstrated to cross-react with rat and mouse pancreatic acinar cells.
Specifications
| PTF1A | |
| Monoclonal | |
| 0.5mg/mL | |
| PTF1α; Ptf1a; Ptf1α; bHLHa29; PTF1-p48 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse, Rat | |
| IgG1 κ |
| Flow Cytometry | |
| S25-763 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human PTF1A Recombinant Protein | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.