Learn More
PTEN Mouse, PE, Clone: A2B1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560002
Description
Cancer can develop when cells escape normal growth control mechanisms through mutations in proto-oncogenes or tumor suppressor genes. A characteristic of most oncogene and tumor suppressor gene products is that they are components of signal transduction pathways that are essential for maintaining cellular homeostasis. PTEN (phosphatase and tensin homolog), also known as MMAC1 (mutated in multiple advanced cancers 1), is a tumor suppressor gene that is mutated at high frequency in multiple tumor types. The protein encoded by PTEN is a phosphatase that preferentially dephosphorylates phosphoinositide substrates. It is believed that a mechanism by which PTEN mutations cause tumors is the loss of its negative control on the phosphoinositide 3-kinase signaling pathway that regulates cell growth and survival. PTEN also plays a role in the maintenance of hematopoietic stem cells.
The A2B1 monoclonal antibody recognizes PTEN, regardless of phosphorylation status.
Specifications
| PTEN | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human C-terminal PTEN Peptide | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| A2B1 | |
| MMAC1, TEP1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Canine, Human, Mouse, Rat | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.