missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PSMD5, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand: Abnova H00005711-AP22.1Set
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-PSMD5 (100μg) , Detection antibody: mouse purified polyclonal anti-PSMD5 (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
| PSMD5 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PSMD5. | |
| KIAA0072/MGC23145/S5B | |
| Mouse, Rabbit | |
| RUO | |
| 5711 | |
| Human | |
| Liquid |
| ELISA | |
| Unconjugated | |
| PSMD5 | |
| PSMD5 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PSMD5 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-PSMD5 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction