missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMB10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544572
This item is not returnable.
View return policy
Description
Peptide sequence: DLTGPQLYGV HPHGSYSRLP FTALGSGQDA ALAVLEDRFQ PNMTLEAAQG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
The 20S proteasome is a protease complex that is responsible for cytosolic protein degradation and generation of peptide ligands for major histocompatibility complex (MHC) class I molecules, either in their final form or in the form of amino-terminally extended precursors. Upon IFN-gamma stimulation of cells, three constitutively expressed subunits of the 20S proteasome are replaced by inducible subunits LMP2 (low-molecular mass polypeptide 2), LMP7, and MECL-1 (multicatalytic endopeptidase complex-like-1, LMP10). LMP2, LMP7, and MECL-1 subunits form immunoproteasomes, which are associated with more efficient class I antigen processing and presentation. Independent assortment of LMP-2, LMP-7, and MECL-1 into different proteasome complexes can lead to 36 unique proteasome subsets, which may mediate differences in the cleavage specificities/cleavage motifs of proteins subject to constitutive- and immuno-proteasomes.
Specifications
| PSMB10 | |
| Polyclonal | |
| Unconjugated | |
| PSMB10 | |
| beta2i; LMP10; Low molecular mass protein 10; macropain; Macropain subunit MECl-1; MECL1; Mecl-1; MGC1665; multicatalytic endopeptidase complex subunit MECl-1; OTTHUMP00000174858; prosome; prosome Mecl1; proteasome (prosome, macropain) subunit, beta type 10; proteasome (prosome, macropain) subunit, beta type, 10; proteasome (prosomome, macropain) subunit, beta type 10; proteasome 20S subunit beta 10; proteasome beta 10 subunit; proteasome catalytic subunit 2i; Proteasome MECl 1; proteasome MECl-1; proteasome subunit 10; proteasome subunit beta 10; proteasome subunit beta 7i; proteasome subunit beta type-10; Proteasome subunit beta-2i; proteasome subunit LMP10; proteasome subunit MECL1; PSMB 10; Psmb10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5699 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P40306 | |
| PSMB10 | |
| Synthetic peptide directed towards the middle region of human PSMB10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering