missing translation for 'onlineSavingsMsg'
Learn More

PSD-95 Mouse, Unlabeled, Clone: 16, BD

Mouse Monoclonal Antibody

£290.95 - £361.00

Specifications

Antigen PSD-95
Clone 16
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15432233
View Documents
BD Biosciences
610495
50 μg
£361.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15855489
View Documents
BD Biosciences
610496
150 μg
ONLINE EXCLUSIVE PRICE
£303.00
£290.95 / 1 set
Save £12.05
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PostSynaptic Density (PSD) refers to a dense region of submembranous cytoskeleton found most prominently in postsynaptic membranes of the CNS. Possible functions for the PSD include regulation and aggregation of receptors,structural stabilization of the synaptic junction, and transduction of signals from membrane receptors. Some of the proteins associated with the PSD are fodrin, tubulin, calmodulin, CaM Kinase II, PSD-95, and PSD-93. PSD-95 (SAP90) is a protein that interacts with the NMDA receptor NMDAR2B, neuronal NOS(nNOS or bNOS), and other proteins. PSD-95 contains one SH3 domain in its carboxy-terminal domain, as well as three conserved repeat regions called GLGF or PDZ domains. nNOS, which is concentrated in synaptic junctions, also contains a PZD domain. PSD-95 and nNOS interact via their respective PZD domains, which may mediate the binding of nNOS to skeletal muscle syntrophin.

This antibody is routinely tested by western blot analysis. Other applications were tested in BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

PSD-95
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
PostSynaptic Density-95; SAP90
16
Western Blot
Unconjugated
RUO
Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.