Learn More
TFF1 Monoclonal Antibody (GE2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512669
Description
MA5-12669 targets pS2/pNR-2 Estrogen-Regulated Protein in IHC (P) applications and shows reactivity with Human samples. The MA5-12669 immunogen is a Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCF-YPNTIDVPPEEECEF (aa 57-84) from C-terminus of human pS2 protein.
Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21.
Specifications
| TFF1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P04155 | |
| TFF1 | |
| A Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCF-YPNTIDVPPEEECEF (aa 57-84) from C-terminus of human pS2 protein | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| GE2 | |
| Unconjugated | |
| TFF1 | |
| Bcei; breast cancer estrogen-inducible protein; breast cancer estrogen-inducible sequence; D21S21; gastrointestinal trefoil protein pS2; HP1.A; HPS2; PNR-2; polypeptide P1.A; Protein pS2; PS2; TFF1; Trefoil factor 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 7031 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.