missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypsin Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524228
This item is not returnable.
View return policy
Description
This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is secreted by the pancreas and cleaved to its active form in the small intestine. It is active on peptide linkages involving the carboxyl group of lysine or arginine. Mutations in this gene are associated with hereditary pancreatitis. This gene and several other trypsinogen genes are localized to the T cell receptor beta locus on chromosome 7.
Specifications
| Trypsin | |
| Polyclonal | |
| PRSS1 | |
| TRP1, TRY1, TRY4, TRYP1, beta-trypsin, cationic trypsinogen, digestive zymogen, nonfunctional trypsin 1, trypsin-1, trypsinogen 1, trypsinogen A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5644 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| P07477 | |
| PRSS1 | |
| KLH conjugated synthetic peptide between 81-107 amino acids from the central region of human PRSS1 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction