missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRPF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541795
This item is not returnable.
View return policy
Description
Peptide sequence: VDKLFEAVEE GRSSRHSKSS SDRSRKRELK EVFGDDSEIS KESSGVKKRR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs. The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs.
Specifications
| PRPF3 | |
| Polyclonal | |
| Unconjugated | |
| PRPF3 | |
| 3632413F13Rik; HPRP3; HPRP3P; MGC137653 protein; pre-mRNA processing factor 3; pre-mRNA-splicing factor 3; PRP3; PRP3 pre-mRNA processing factor 3 homolog; PRP3 pre-mRNA processing factor 3 homolog (yeast); Prp3p; PRPF3; RP18; SNRNP90; U4/U6 small nuclear ribonucleoprotein Prp3; U4/U6 snRNP 90 kDa protein; U4/U6-associated RNA splicing factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9129 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O43395 | |
| PRPF3 | |
| Synthetic peptide directed towards the N-terminal of human PRPF3 (aa 63-112). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction