Learn More
Invitrogen™ Proteus Monoclonal Antibody (31-17)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA135772
Description
The antibody reacts with bacteria in suspension and is therefore useful for detection of Proteus spp. in bacterial suspensions. Various systems can be used with this antibody to detect the presence of Proteus spp.. The antibody reacts with approximately 50% of the Proteus spp. isolates tested.
Specifications
| Proteus | |
| Monoclonal | |
| 0.10 mg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| Proprietary immunogen. | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA | |
| 31-17 | |
| Unconjugated | |
| Mouse | |
| Purified | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.