Learn More
Invitrogen™ PR3 Monoclonal Antibody (3B4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515540
Description
MA5-15540 targets Proteinase 3 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15540 immunogen is purified recombinant fragment of human Proteinase 3 expressed in E. Coli. MA5-15540 detects Proteinase 3 which has a predicted molecular weight of approximately 28kDa.
Serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) (PubMed:3198760, PubMed:2033050, PubMed:28240246). By cleaving and activating receptor F2RL1/PAR-2, enhances endothelial cell barrier function and thus vascular integrity during neutrophil transendothelial migration (PubMed:23202369). May play a role in neutrophil transendothelial migration, probably when associated with CD177 (PubMed:22266279). [UniProt].
Specifications
| PR3 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P24158 | |
| PRTN3 | |
| Purified recombinant fragment of human Proteinase 3 expressed in E. Coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 3B4 | |
| Unconjugated | |
| PRTN3 | |
| ACPA; AGP7; azurophil granule protein 7; CANCA; C-ANCA; C-ANCA antigen; leukocyte proteinase 3; MBN; MBT; mPR3; Myeloblastin; neutrophil proteinase 4; NP4; NP-4; P29; PR3; PR-3; Proteinase 3; PRTN3; serine proteinase, neutrophil; wegener autoantigen; Wegener granulomatosis autoantigen | |
| Mouse | |
| Protein G | |
| RUO | |
| 5657 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.