missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R5D Polyclonal Antibody

Product Code. 11575942
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11575942 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11575942 Supplier Invitrogen™ Supplier No. PA112389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPP2R5D Polyclonal Antibody for Western Blot

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZNF265 (Zinc finger protein 265), also known as ZRANB2 (Zinc finger Ran-binding domain-containing protein 2), ZIS, ZIS1 or ZIS2, is a 330 amino acid protein that belongs to the ZRANB2 family. Localized to the nucleus, ZNF265 functions as a splicing factor that is responsible for alternatively splicing Tra-2 beta (transformer-2 beta) transcripts and is thought to interfere with constitutive 5'-splice selection. ZNF265 contains two RanBP2-type zinc fingers through which it conveys its RNA-binding activity. Two isoforms, designated ZIS-1 and ZIS-2, are expressed due to alternative splicing events. Upon DNA damage, ZIS-2 may be phosphorylated by ATM or ATR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PPP2R5D
Gene Accession No. Q14738
Gene Alias B56D; B'delta; MGC2134; MGC8949; MRD35; PP2A B subunit isoform B56-delta; PP2A B subunit isoform B'-delta; PP2A B subunit isoform PR61-delta; PP2A B subunit isoform R5-delta; PP2A, B subunit, B' delta isoform; PP2A, B subunit, B56 delta isoform; PP2A, B subunit, PR61 delta isoform; PP2A, B subunit, R5 delta isoform; PP2CB; Ppp2r5d; protein phosphatase 2 regulatory subunit B'delta; protein phosphatase 2, regulatory subunit B (B56), delta isoform; protein phosphatase 2, regulatory subunit B', delta; protein phosphatase 2, regulatory subunit B', delta isoform; Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, delta isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform; TEG-271; testis expressed gene 271; Tex271
Gene Symbols PPP2R5D
Host Species Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to the N-terminus of PP2A/B delta.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21770, 363193, 5528
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.