missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R5A Polyclonal Antibody

Product Code. 11555942
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11555942 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11555942 Supplier Invitrogen™ Supplier No. PA112387

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PPP2R5A Polyclonal Antibody for Western Blot, IHC

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B56 subfamily. Alternative transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PPP2R5A
Gene Accession No. Q15172, Q6PD03
Gene Alias B56A; MGC131915; PP2A B subunit isoform B56-alpha; PP2A B subunit isoform B'-alpha; PP2A B subunit isoform PR61-alpha; PP2A B subunit isoform R5-alpha; PP2A, B subunit, B' alpha isoform; PP2A, B subunit, B56 alpha isoform; PP2A, B subunit, PR61 alpha isoform; PP2A, B subunit, R5 alpha isoform; PP2CB; PPP2R5A; PR61A; PR61alpha; protein phosphatase 2 regulatory subunit B', alpha; protein phosphatase 2 regulatory subunit B'alpha; protein phosphatase 2, regulatory subunit B (B56), alpha isoform; protein phosphatase 2, regulatory subunit B', alpha; protein phosphatase 2, regulatory subunit B', alpha isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alpha isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform
Gene Symbols PPP2R5A
Host Species Sheep
Immunogen Synthetic peptide.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226849, 312754, 5525
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.