Learn More
Protein A Goat anti-Bacteria, HRP, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA126853
Description
PA1-26853 detects Protein A. PA1-26853 has been successfully used in immunohistochemistry (frozen), ELISA, and Western blot procedures. The PA1-26853 immunogen is protein A from Staphylococcus aureus. Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20° C or below, avoiding multiple freeze-thaw cycles.
Protein A is 56 kDA surface protein found in the cell wall of Staphylococcus aureus. Protein A has a high affinity to Human IgG, Mouse IgG and others and is therefore suitable for purification and detection of immunglobulins.
Specifications
| Protein A | |
| Polyclonal | |
| HRP | |
| Protein A from Staphylococcus aureus. | |
| 1 mg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Western Blot | |
| 10 mg/mL | |
| Goat | |
| IgG fraction | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.