missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prostein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146229
This item is not returnable.
View return policy
Description
Prostein Polyclonal Antibody for Western Blot, IHC (P), IHC (F), ICC/IF, ELISA
Prostein is a novel prostate-specific protein expressed in normal and malignant prostate tissues. This protein appears to be a type IIIa plasma membrane protein with a cleavable signal peptide and 11 transmembrane-spanning regions. Prostein expression is androgen responsive since treatment of LNCaP cells with androgen up-regulates prostein message and protein expression levels. Thus, prostein is a prostate-specific marker whose detection and quantitation may be useful in diagnosing and monitoring prostate cancer.
Specifications
| Prostein | |
| Polyclonal | |
| Unconjugated | |
| SLC45A3 | |
| 2210413P12Rik; AU023994; AU043764; IPCA-2; IPCA6; IPCA-6; IPCA-8; PCANAP2; PCANAP6; PCANAP8; prostate cancer associated protein 2; prostate cancer associated protein 6; prostate cancer associated protein 8; prostate cancer-associated gene 2; prostate cancer-associated gene 6; prostate cancer-associated gene 8; prostate cancer-associated protein 6; Prostein; PRST; RGD1309764; SLC45A3; Solute carrier family 45 member 3; solute carrier family 45, member 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85414 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q96JT2 | |
| SLC45A3 | |
| Synthetic peptides corresponding to the N terminal sequence and the C terminal sequence of the human Prostein protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction