missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Procalcitonin Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA175362
This item is not returnable.
View return policy
Description
Procalcitonin Polyclonal Antibody for ELISA
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| Procalcitonin | |
| Polyclonal | |
| Unconjugated | |
| CALCA | |
| alpha-type CGRP; CALC1; CALCA; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCP; CGRP; CGRP1; CGRP-I; CT; Katacalcin; KC; PCT; PDN-21 | |
| Sheep | |
| Affinity chromatography | |
| RUO | |
| 796 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| 2.659 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01258, P06881 | |
| CALCA | |
| Recombinant human PCT (aa 3-116). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction