Learn More
Pre-B Cell Receptor Rat anti-Mouse, Biotin, Clone: SL156, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 551863
Description
The pre-B cell receptor (pre-BCR) expressed during the early stages of B lymphocyte development is a heterodimer of immunoglobulin heavy chain (IgH) with surrogate light chain, which is an Ig-light-chain-like molecule composed of the non-covalently linked Lambda 5 (λ5) and VpreB proteins. The pre-BCR is believed to control IgH repertoire selection and proliferation of differentiating B lymphocytes. The SL156 antibody reacts with a conformational epitope of the pre-BCR in transfected X63-Ag8.653 cells but not with surrogate light chain, λ5, or VpreB in the absence of IgH. It detects pre-BCR on pre-B cell lines, but not on pro-B cell lines or IgM-positive splenocytes. It also detects pre-BCR on the surface of pre-B cells, but not on Ig light chain (IgL)-positive B lymphocytes, from the bone marrow of normal mice. It has been noted that the cell-surface expression of pre-BCR is upregulated after a one-hour incubation of bone-marrow leukocytes in tissue culture medium at 37°C. After immunization, cell-surface pre-BCR is detected on a subset of splenic IgL+ germinal-center B lymphocytes.
Host Species: Rat
Clone: SL156
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Purified soluble complex of recombinant μ IgH, λ5, and VpreB
Flow Cytometry
Specifications
| Pre-B Cell Receptor | |
| Monoclonal | |
| 0.5mg/mL | |
| Pre-BCR | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| SL156 | |
| Biotin | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Purified soluble complex of recombinant μ IgH, λ5, and VpreB | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.