Learn More
anti-PRDX3 Matched Antibody Pair, Ab-1, Abnova
Brand: Abnova H00010935-AP11.1Set
Additional Details : Weight : 0.00010kg
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-PRDX3 (100μg); Mouse monoclonal anti-PRDX3, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This gene encodes a protein with antioxidant function and is localized in the mitochondrion. This gene shows significant nucleotide sequence similarity to the gene coding for the C22 subunit of Salmonella typhimurium alkylhydroperoxide reductase. Expression of this gene product in E. coli deficient in the C22-subunit gene rescued resistance of the bacteria to alkylhydroperoxide. The human and mouse genes are highly conserved, and they map to the regions syntenic between mouse and human chromosomes. Sequence comparisons with recently cloned mammalian homologues suggest that these genes consist of a family that is responsible for regulation of cellular proliferation, differentiation, and antioxidant functions. Two transcript variants encoding two different isoforms have been found for this gene.
ELISA
Specifications
PRDX3 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PRDX3. | |
AOP-1/AOP1/MER5/MGC104387/MGC24293/PRO1748/SP-22 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PRDX3 (100 μg) 2. Detection antibody: mouse monoclonal anti-PRDX3, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
PRDX3 | |
PRDX3 | |
Affinity Purified | |
RUO | |
10935 | |
Human | |
IgG1 |