missing translation for 'onlineSavingsMsg'
Learn More

anti-PPP2R3B Ab-3 Matched Antibody Pair, Abnova™

Brand:  Abnova H00028227-AP21.1Set

Product Code. 16193334

  • £729.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!


Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PPP2R3B (100μg); Detection antibody: mouse purified polyclonal anti-PPP2R3B (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
This item is not returnable. View return policy

Description

Description

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PPP2R3B

Host Species: Rabbit, Mouse
Species Reactivity: Hu

ELISA Pair

Specifications

Specifications

PPP2R3B
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PPP2R3B.
NY-REN-8/PPP2R3L/PPP2R3LY/PR48
Mouse, Rabbit
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PPP2R3B (100 μg) 2. Detection antibody: mouse purified polyclonal anti-PPP2R3B (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
Unconjugated
PPP2R3B
PPP2R3B
Affinity Purified
RUO
28227
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.