missing translation for 'onlineSavingsMsg'
Learn More

PP2A Catalytic α Mouse, Unlabeled, Clone: 46, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610555

 View more versions of this product

Product Code. 15825519

  • ONLINE EXCLUSIVE PRICE
    £229.00
    £219.86 / 1 set
    Save £9.14
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

P rotein p hosphatases of the type 2A (PP2A) are composed of two regulatory and one catalytic subunit. The different subunits are encoded by multiple genes. PP2A are specific for phosphoserine and phosphothreonine residues of proteins that are involved in the regulation of cellular growth. PP2A binds and dephosphorylates the middle T antigens of polyoma virus. This interaction may regulate the transcriptional activity of the virus. The 36 kDa catalytic subunit is rapidly tyrosine phosphorylated as a result of growth factor stimulation or cell transformation. Tyrosine phosphorylation of the catalytic subunit may inactivate the phosphatase activity of PP2A.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PP2A Catalytic α
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Western Blot
46
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.