missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R2A Monoclonal Antibody (F.722.1), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA515007
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the B-prime (PR61), B-prime-prime or B-prime-prime-prime families of PP2A B subunits.
The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B55 subfamily. Alternatively spliced transcript variants have been described.Specifications
PPP2R2A | |
Monoclonal | |
104 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P36876, P63151, Q6P1F6 | |
PPP2R2A | |
Synthetic peptide corresponding to the sequence of human PP2A B subunit | |
100 μL | |
Primary | |
Drosophila, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
F.722.1 | |
Unconjugated | |
PPP2R2A | |
2410004D02Rik; B55A; B55ALPHA; hypothetical protein; mKIAA1541; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform BRA; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform R2-alpha; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, R2-alpha isoform; PPP2R2A; PR52A; PR55A; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2 regulatory subunit B, alpha; protein phosphatase 2 regulatory subunit Balpha; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2A 55 kDa regulatory subunit, alpha isoform; serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alpha isoform; serine/threonine-protein phosphatase 2A 55 | |
Rabbit | |
Affinity chromatography | |
RUO | |
117104, 5520, 71978 | |
-20°C | |
Liquid |