missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534469
Code : NEW
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is NIH-3T3 cell lysate. PA5-34469 can be used with blocking peptide PEP-1511.
POU3F2 Polyclonal antibody specifically detects POU3F2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
POU3F2 | |
Polyclonal | |
Unconjugated | |
POU3F2 | |
7-Oct; 9430075J19Rik; A230098E07Rik; Brain-2; brain-2 class III POU-domain protein; brain-specific homeobox/POU domain protein 2; BRN2; Brn-2; Nervous system-specific octamer-binding transcription factor N-Oct-3; N-Oct3; OCT7; Oct-7; Octamer-binding protein 7; octamer-binding transcription factor 7; Otf7; OTF-7; POU class 3 homeobox 2; POU domain, class 3, transcription factor 2; POU3F2; POUF3; protein Brn-2; Unknown (protein for MGC:137739) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18992, 29588, 5454 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P20265, P31360, P56222 | |
POU3F2 | |
An 18 amino acid peptide near the carboxy terminus of human POU3F2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |