missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-POLR1D, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00051082-PW1.1Set
Includes: Antibody pair for IP: rabbit polyclonal anti-POLR1D (300μL), Antibody pair for WB: rabbit polyclonal anti-POLR1D (50μL)
Spécification
| POLR1D | |
| Monoclonal/Polyclonal | |
| This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
| FLJ20616/MGC9850/POLR1C/RPA16/RPA9/RPAC2/RPO1-3 | |
| Rabbit | |
| RUO | |
| 51082 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| POLR1D | |
| POLR1D | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-POLR1D (300 ul) 2. Antibody pair for WB: rabbit polyclonal anti-POLR1D (50 ul) | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu