missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phospholipid scramblase 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PLSCR1 protein.
Brand: Abnova H00005359-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MDKQNSQMNASHPETNLPVGYPPQYPPTAFQGPPGYSGYPGPQVSYPPPPAGHSGPGPAGFPVPNQPVYNQPVYNQPVGAAGVPWMPAPQPPLNCPPGLEYLSQIDQILIHQQIELLEVLTGFETNNKYEIKNSFGQRVYFAAEDTDCCTRNCCGPSRPFTLRIIDNMGQEVITLERPLRCSSCCCPCCLQEIEIQAPPGVPIGYVIQTWHPCLPKFTIQNEKREDVLKISGPCVVCSCCGDVDFEIKSLDEQCVVGKISKHWTGILREAFTDADNFGIQFPLDLDVKMKAVMIGACFLIDFMFFESTGSQEQKSGVWSpecifications
| phospholipid scramblase 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human PLSCR1 protein. | |
| PLSCR1 | |
| MMTRA1B | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 5359 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_021105.1 | |
| PLSCR1 | |
| PLSCR1 (NP_066928.1, 1 a.a. ∼ 318 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction