Learn More
PLXNC1 Goat anti-Human, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547549
Description
In direct ELISAs and Western blots, approximately10% cross-reactivity with recombit mouse (rm) Plexin A1, rmPlexin A2, rmPlexin A3, and recombit human (rh) Plexin B1 is observed, and less than 1% cross-reactivity with rhPlexin D1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
| PLXNC1 | |
| Polyclonal | |
| Unconjugated | |
| PLXNC1 | |
| CD232, PLXNC1, VESPR, CD232, CD232 antigen, plexin (semaphorin receptor), plexin C1, PLXN-C1, receptor for viral semaphorin protein, receptor for virally-encoded semaphorin, VESPRplexin-C1, Virus-encoded semaphorin protein receptor | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 10154 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O60486 | |
| PLXNC1 | |
| E. coli-derived recombit human Plexin C1 Ile49-Ser161 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.