Learn More
pleckstrin homology domain containing, family M (with RUN domain) member 1, Mouse, Clone: 1C9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PLEKHM1.
Brand: Abnova H00009842-M01.100ug
Description
The protein encoded by this gene is essential for bone resorption, and may play a critical role in vesicular transport in the osteoclast. Mutations in this gene are associated with autosomal recessive osteopetrosis type 6 (OPTB6). Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Sequence: PHRFSVADLQQIADGVYEGFLKALIEFASQHVYHCDLCTQRGFICQICQHHDIIFPFEFDTTVRCAECKTVFHQSCQAVVKKGCPRCARRRKYQEQNIFASpecifications
| pleckstrin homology domain containing, family M (with RUN domain) member 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC064361 | |
| PLEKHM1 | |
| PLEKHM1 (AAH64361, 957 a.a. ∼ 1056 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1C9 | |
| Mouse monoclonal antibody raised against a partial recombinant PLEKHM1. | |
| PLEKHM1 | |
| AP162/B2/KIAA0356/OPTB6 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 9842 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.