missing translation for 'onlineSavingsMsg'
Learn More

fermitin family homolog 2 (Drosophila), Mouse, Clone: 2G11, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant PLEKHC1.

Brand:  Abnova H00010979-M09.100ug

Product Code. 16155952

  • £303.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MALDGIRMPDGCYADGTWELSVHVTDLNRDVTLRVTGEVHIGGVMLKLVEKLDVKKDWSDHALWWEKKRTWLLKTHWTLDKYGIQADAKLQFTPQHKLLRLQLPNMKYVKVKVNFSDRVFKAVSDICKTFNIRHPEELSLLKKPRDPTKKKKKKLDDQSEDEALELEGPLITPGSGSIYSSPGLYSKTMTPTYDAHDGSPLSPTSAWFGDSALSEGNPGILAVSQPITSPEILAKMFKPQALLDKAKINQGWLDSSRSLMEQDVKENEALLLRFKYYSFFDLNPKYDAIRINQLYEQAKWAILLEEIECTEEEMMMFAALQYHINKLSIMTSENHLNNSDKEVDEVDAALSDLEITLEGGKTSTILGDITSIPELADYIKVFKPKKLTLKGYKQYWCTFKDTSISCYKSKEESSGTPAHQMNLRGCEVTPDVNISGQKFNIKLLIPVAEGMNEIWLRCDNEKQYAHWMAACRLASKGKTMADSSYNLEVQNILSSLKMQHLNPDPQLIPEQITTDITPECLVSPRYLKKYKNKQITARILEAHQNVAQMSLIEAKMRFIQAWQSLPEFGITHFIARFQGGKKEELIGIAYNRLIRMDASTGDAIKTWRFSNMKQWNVNWEIKMVTVEFADEVRLSFICTEVDCKVVHEFIGGYIFLSTRAKDQNESLDEEMFYKLTSGWV
Specifications

Specifications

fermitin family homolog 2 (Drosophila)
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
BC017327
FERMT2
PLEKHC1 (AAH17327, 1 a.a. ∼ 680 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Cell Adhesion
10979
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG2a κ
ELISA, Immunofluorescence, Western Blot
2G11
Mouse monoclonal antibody raised against a full length recombinant PLEKHC1.
FERMT2
DKFZp686G11125/FLJ34213/FLJ44462/KIND2/MIG2/PLEKHC1/UNC112/UNC112B/mig-2
Mouse
Affinity Purified
RUO
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.